missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydrobenzo[b]furan-5-carbaldehyde, 95%, Maybridge

Chemical Identifiers

CAS 55745-70-5
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00068058
InChI Key WEBVDBDZSOJGPB-UHFFFAOYSA-N
Synonym 2,3-dihydro-5-benzofurancarbaldehyde
PubChem CID 735901
IUPAC Name 2,3-dihydro-1-benzofuran-5-carbaldehyde
SMILES C1COC2=C1C=C(C=C2)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC00804DA SDS View Documents Fisher Scientific Company
CC00804DA
1g Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

55745-70-5
148.161
WEBVDBDZSOJGPB-UHFFFAOYSA-N
735901
C1COC2=C1C=C(C=C2)C=O
C9H8O2
MFCD00068058
2,3-dihydro-5-benzofurancarbaldehyde
2,3-dihydro-1-benzofuran-5-carbaldehyde

Specifications

Amber Glass Bottle
55745-70-5
100.0
C9H8O2
2,3-dihydro-5-benzofurancarbaldehyde
C1COC2=C1C=C(C=C2)C=O
148.161
148.16
2, 3-Dihydrobenzo[b]furan-5-carbaldehyde
1g
94.5
95%
MFCD00068058
WEBVDBDZSOJGPB-UHFFFAOYSA-N
2,3-dihydro-1-benzofuran-5-carbaldehyde
735901
95%