Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dihydro-6-phenylimidazo[2,1-b]thiazole-5-carboxaldehyde, 97%

$127.91 - $557.22

Chemical Identifiers

Chemical Name or Material 2,3-Dihydro-6-phenylimidazo[2,1-b]thiazole-5-carboxaldehyde
CAS 75224-64-5
Molecular Formula C12H10N2OS
Formula Weight 230.29
MDL Number MFCD00268936
Synonym 6-Phenyl-2,3-dihydroimidazo[2,1-b]thiazole-5-carboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5048203

 
alfa aesar™
H5048203
1g Each for $127.91

AAH5048206

 
alfa aesar™
H5048206
5g Each for $557.22
Description & Specifications

Specifications

Formula Weight 230.29
Chemical Name or Material 2,3-Dihydro-6-phenylimidazo[2,1-b]thiazole-5-carboxaldehyde
Assay 97%
Molecular Formula C12H10N2OS
MDL Number MFCD00268936
Melting Point 122° to 124°C
CAS 75224-64-5
Synonym 6-Phenyl-2,3-dihydroimidazo[2,1-b]thiazole-5-carboxaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.