Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dihydro-5H-1,4-benzodioxepin-5-one, 98%

$110.14 - $406.82

Chemical Identifiers

Chemical Name or Material 2,3-Dihydro-5H-1,4-benzodioxepin-5-one
CAS 22891-52-7
Molecular Formula C9H8O3
Formula Weight 164.16
MDL Number MFCD00236035
Beilstein 4464
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1358203

  SDS
alfa aesar™
L1358203
1g Each for $110.14

AAL1358206

  SDS
alfa aesar™
L1358206
5g Each for $406.82
Description & Specifications

Specifications

Formula Weight 164.16
Chemical Name or Material 2,3-Dihydro-5H-1,4-benzodioxepin-5-one
Assay 98%
Molecular Formula C9H8O3
MDL Number MFCD00236035
CAS 22891-52-7
Melting Point 29° to 31°C
Beilstein 4464
RTECS Number DF4725800
Boiling Point 178° to 184°C (15mmHg)
Quantity 1g