missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dihydro-5H-1,4-benzodioxepin-5-one, 98%

Manufacturer:  Alfa Aesar™ L1358203

 View more versions of this product

Catalog No. AAL1358203



Specifications

Specifications

2,3-Dihydro-5H-1,4-benzodioxepin-5-one
29°C to 31°C
22891-52-7
MFCD00236035
2,3-dihydro-5h-1,4-benzodioxepin-5-one, 2h-benzo e 1,4 dioxepin-5 3h-one, 2,3-dihydro-5h-1,4-benzodioxipin-5-one, 5h-1,4-benzodioxepin-5-one, 2,3-dihydro, 4-19-00-01699 beilstein handbook reference, 2h,3h-benzo f 1,4-dioxepin-5-one
C1COC(=O)C2=CC=CC=C2O1
164.16
164.16
178°C to 184°C (15mmHg)
1g
C9H8O3
4464
PYVNMBGWXYJCCL-UHFFFAOYSA-N
2,3-dihydro-1,4-benzodioxepin-5-one
211314
98%

Chemical Identifiers

22891-52-7
164.16
PYVNMBGWXYJCCL-UHFFFAOYSA-N
211314
C1COC(=O)C2=CC=CC=C2O1
C9H8O3
MFCD00236035
2,3-dihydro-5h-1,4-benzodioxepin-5-one, 2h-benzo e 1,4 dioxepin-5 3h-one, 2,3-dihydro-5h-1,4-benzodioxipin-5-one, 5h-1,4-benzodioxepin-5-one, 2,3-dihydro, 4-19-00-01699 beilstein handbook reference, 2h,3h-benzo f 1,4-dioxepin-5-one
2,3-dihydro-1,4-benzodioxepin-5-one
Safety and Handling

Safety and Handling

RTECSNumber : DF4725800

Documents