missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dihydro-2,2-dimethyl-7-hydroxybenzofuran 98.0+%, TCI America™
$64.11 - $320.53
Chemical Identifiers
CAS | 1563-38-8 |
---|---|
Molecular Formula | C10H12O2 |
Molecular Weight (g/mol) | 164.204 |
MDL Number | MFCD00075382 |
InChI Key | WJGPNUBJBMCRQH-UHFFFAOYSA-N |
Synonym | 2,2-Dimethyl-7-hydroxycoumaran |
PubChem CID | 15278 |
ChEBI | CHEBI:38474 |
IUPAC Name | 2,2-dimethyl-3H-1-benzofuran-7-ol |
SMILES | CC1(CC2=C(O1)C(=CC=C2)O)C |
Chemical Identifiers
1563-38-8 | |
164.204 | |
WJGPNUBJBMCRQH-UHFFFAOYSA-N | |
15278 | |
2,2-dimethyl-3H-1-benzofuran-7-ol |
C10H12O2 | |
MFCD00075382 | |
2,2-Dimethyl-7-hydroxycoumaran | |
CHEBI:38474 | |
CC1(CC2=C(O1)C(=CC=C2)O)C |
Specifications
95°C | |
25g | |
C10H12O2 | |
2,2-Dimethyl-7-hydroxycoumaran | |
CC1(CC2=C(O1)C(=CC=C2)O)C | |
164.204 | |
CHEBI:38474 | |
Clear Liquid at 20°C | |
2,3-Dihydro-2,2-dimethyl-7-hydroxybenzofuran |
Yellow | |
1563-38-8 | |
MFCD00075382 | |
WJGPNUBJBMCRQH-UHFFFAOYSA-N | |
2,2-dimethyl-3H-1-benzofuran-7-ol | |
15278 | |
164.20 | |
≥98.0% (GC) |
Safety and Handling
EINECSNumber : (5)-5541
TSCA : Yes