missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydro-1-benzofuran-7-ylmethylamine, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC00913DA

 View more versions of this product

Catalog No. CC00913DA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 3-Dihydro-1-benzofuran-7-ylmethylamine
361393-65-9
100.0
97%
C9H11NO
MFCD11109312
1g
96.5
Amber Glass Bottle
97%
149.19

Chemical Identifiers

361393-65-9
100.0
C9H11NO
MFCD11109312
97%
96.5
97%
149.19
2, 3-Dihydro-1-benzofuran-7-ylmethylamine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger