missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydro-1-benzofuran-7-ylmethylamine, 97%, Maybridge

$114.15 - $114.15

Chemical Identifiers

CAS 361393-65-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H11NO
Formula Weight 149.19
MDL Number MFCD11109312
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-7-ylmethylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC00913CB SDS Fisher Scientific Company
CC00913CB
Amber Glass Bottle 250mg Each for $114.15
CC00913DA SDS Fisher Scientific Company
CC00913DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

361393-65-9
100.0
C9H11NO
MFCD11109312
97%
96.5
97%
149.19
2, 3-Dihydro-1-benzofuran-7-ylmethylamine

Specifications

250mg
96.5
Amber Glass Bottle
C9H11NO
MFCD11109312
97%
361393-65-9
100.0
97%
149.19
2, 3-Dihydro-1-benzofuran-7-ylmethylamine

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger