Please login to your online account to display your discounted pricing

2,3-Dihydro-1-benzofuran-7-sulfonyl chloride, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC00903CB

 View more versions of this product

Catalog No. CC00903CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 218.66
Quantity 250mg
Assay 97%
CAS 953408-82-7
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-7-sulfonyl chloride
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C8H7ClO3S
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 2, 3-Dihydro-1-benzofuran-7-sulfonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H7ClO3S
Formula Weight 218.66
Accessories

GHS Signal Word: Danger