missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydro-1-benzofuran-7-sulfonyl chloride, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC00903DE

 View more versions of this product

Catalog No. CC00903DE

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

218.66
97%
Amber Glass Bottle
97%
5g
2, 3-Dihydro-1-benzofuran-7-sulfonyl chloride
953408-82-7
C8H7ClO3S

Chemical Identifiers

97%
218.66
97%
C8H7ClO3S
2, 3-Dihydro-1-benzofuran-7-sulfonyl chloride

Safety and Handling

SDS & Safety Documents

GHS Signal Word: Danger