missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydro-1-benzofuran-5-yl isocyanate, 97%, Maybridge

Manufacturer:  Thermo Scientific CC00806DE

 View more versions of this product

Catalog No. CC00806DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 3-Dihydro-1-benzofuran-5-yl isocyanate
215162-92-8
100.0
C9H7NO2
2,3-dihydro-1-benzofuran-5-yl isocyanate, 5-isocyanato-2,3-dihydrobenzofuran, 2,3-dihydro-1-benzofuran-5-ylisocyanate, 2,3-dihydrobenzo b furan-5-isocyanate, benzofuran,2,3-dihydro-5-isocyanato, 2,3-dihydrobenzofuran-5-yl isocyanate, 2,3-dihydro-5-isocyanatobenzo b furan, 5-isocyanato-2,3-dihydrobenzo b furan, 2,3-dihydro-5-isocyanato-1-benzofuran, 2,3-dihydro-1-benzofuran-5-yl-isocyanate
C1COC2=C1C=C(C=C2)N=C=O
161.16
161.16
5g
96.5
97%
MFCD02677718
WAIKTAFMGLVRJC-UHFFFAOYSA-N
5-isocyanato-2,3-dihydro-1-benzofuran
2776156
97%

Chemical Identifiers

215162-92-8
161.16
WAIKTAFMGLVRJC-UHFFFAOYSA-N
2776156
C1COC2=C1C=C(C=C2)N=C=O
C9H7NO2
MFCD02677718
2,3-dihydro-1-benzofuran-5-yl isocyanate, 5-isocyanato-2,3-dihydrobenzofuran, 2,3-dihydro-1-benzofuran-5-ylisocyanate, 2,3-dihydrobenzo b furan-5-isocyanate, benzofuran,2,3-dihydro-5-isocyanato, 2,3-dihydrobenzofuran-5-yl isocyanate, 2,3-dihydro-5-isocyanatobenzo b furan, 5-isocyanato-2,3-dihydrobenzo b furan, 2,3-dihydro-5-isocyanato-1-benzofuran, 2,3-dihydro-1-benzofuran-5-yl-isocyanate
5-isocyanato-2,3-dihydro-1-benzofuran
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Documents