missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydro-1-benzofuran-2-carboxylic acid, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC23901EA

 View more versions of this product

Catalog No. CC23901EA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 3-Dihydro-1-benzofuran-2-carboxylic acid
1914-60-9
100.0
97%
C9H8O3
MFCD03086168
10g
96.5
Amber Glass Bottle
97%
164.16

Chemical Identifiers

1914-60-9
100.0
C9H8O3
MFCD03086168
97%
96.5
97%
164.16
2, 3-Dihydro-1-benzofuran-2-carboxylic acid

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning