missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydro-1-benzofuran-2-carboxylic acid, 97%, Thermo Scientific™

$192.93 - $192.93

Chemical Identifiers

CAS 1914-60-9
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD03086168
InChI Key WEVFUSSJCGAVOH-UHFFFAOYNA-N
Synonym 2,3-dihydro-2-benzofurancarboxylic acid, 2,3-dihydro-benzofuran-2-carboxylic acid, 2,3-dihydrobenzo b furan-2-carboxylic acid, 2,3-dihydrobenzofuran carboxylic acid, 2,3-dihydrobenzofuran-2-carboxylic, 2,3-dihydrobenzofuran-2-carboxylic acid, 2,3-dihydrobenzofuran-2-carboxylicacid, 2-benzofurancarboxylic acid, 2,3-dihydro, 2-benzofurancarboxylicacid, 2,3-dihydro, dihydrobenzofuran-2-carboxylic acid
PubChem CID 2776555
IUPAC Name 2,3-dihydro-1-benzofuran-2-carboxylic acid
SMILES OC(=O)C1CC2=CC=CC=C2O1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC23901DA
SDS View Documents Promotion Details
Fisher Scientific Company
CC23901DA
1g Amber Glass Bottle
Each for $192.93
 
Description

Description

Specifications

Chemical Identifiers

1914-60-9
164.16
WEVFUSSJCGAVOH-UHFFFAOYNA-N
2776555
OC(=O)C1CC2=CC=CC=C2O1
C9H8O3
MFCD03086168
2,3-dihydro-2-benzofurancarboxylic acid, 2,3-dihydro-benzofuran-2-carboxylic acid, 2,3-dihydrobenzo b furan-2-carboxylic acid, 2,3-dihydrobenzofuran carboxylic acid, 2,3-dihydrobenzofuran-2-carboxylic, 2,3-dihydrobenzofuran-2-carboxylic acid, 2,3-dihydrobenzofuran-2-carboxylicacid, 2-benzofurancarboxylic acid, 2,3-dihydro, 2-benzofurancarboxylicacid, 2,3-dihydro, dihydrobenzofuran-2-carboxylic acid
2,3-dihydro-1-benzofuran-2-carboxylic acid

Specifications

1914-60-9
100.0
C9H8O3
Amber Glass Bottle
2,3-dihydro-2-benzofurancarboxylic acid, 2,3-dihydro-benzofuran-2-carboxylic acid, 2,3-dihydrobenzo b furan-2-carboxylic acid, 2,3-dihydrobenzofuran carboxylic acid, 2,3-dihydrobenzofuran-2-carboxylic, 2,3-dihydrobenzofuran-2-carboxylic acid, 2,3-dihydrobenzofuran-2-carboxylicacid, 2-benzofurancarboxylic acid, 2,3-dihydro, 2-benzofurancarboxylicacid, 2,3-dihydro, dihydrobenzofuran-2-carboxylic acid
OC(=O)C1CC2=CC=CC=C2O1
164.16
164.16
2, 3-Dihydro-1-benzofuran-2-carboxylic acid
96.5
97%
MFCD03086168
1g
WEVFUSSJCGAVOH-UHFFFAOYNA-N
2,3-dihydro-1-benzofuran-2-carboxylic acid
2776555
97%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Product Certifications