Please login to your online account to display your discounted pricing

2,3-Dihydro-1-benzofuran-2-carboxylic acid, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC23901DA

 View more versions of this product

Catalog No. CC23901DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 164.16
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-2-carboxylic acid
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C9H8O3
CAS 1914-60-9
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD03086168
CAS Min % 96.5
Health Hazard 1 Warning

Specifications

Chemical Name or Material 2, 3-Dihydro-1-benzofuran-2-carboxylic acid
CAS 1914-60-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8O3
Formula Weight 164.16
MDL Number MFCD03086168
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.