missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydro-1,4-benzodioxine-6-sulfonyl chloride, 97%, Maybridge™

Chemical Identifiers

CAS 63758-12-3
Molecular Formula C8H7ClO4S
Molecular Weight (g/mol) 234.65
MDL Number MFCD02677681
InChI Key JWHSRWUHRLYAPM-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-6-sulfonyl chloride, 1,4-benzodioxin-6-sulfonyl chloride, 2,3-dihydro, 1,4-benzodioxin-6-sulfonylchloride, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxine-6-sulphonylchloride, 2,3-dihydro-benzo 1,4 dioxine-6-sulfonyl chloride, 2,3-dihydro-benzo 1,4 dioxine-6-sulfonylchloride, 2,3-dihydrobenzo b 1,4 dioxine-6-sulfonyl chloride, 2h,3h-benzo 3,4-e 1,4-dioxin-6-ylchlorosulfone, 6-chlorosulfonyl-benzo-1,4-dioxane, 6-chlorosulphonyl-2,3-dihydro-1,4-benzodioxine
PubChem CID 2776169
IUPAC Name 2,3-dihydro-1,4-benzodioxine-6-sulfonyl chloride
SMILES C1COC2=C(O1)C=CC(=C2)S(=O)(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC01303DA
SDS View Documents
Fisher Scientific Company
CC01303DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

63758-12-3
234.65
JWHSRWUHRLYAPM-UHFFFAOYSA-N
2776169
C1COC2=C(O1)C=CC(=C2)S(=O)(=O)Cl
C8H7ClO4S
MFCD02677681
1,4-benzodioxan-6-sulfonyl chloride, 1,4-benzodioxin-6-sulfonyl chloride, 2,3-dihydro, 1,4-benzodioxin-6-sulfonylchloride, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxine-6-sulphonylchloride, 2,3-dihydro-benzo 1,4 dioxine-6-sulfonyl chloride, 2,3-dihydro-benzo 1,4 dioxine-6-sulfonylchloride, 2,3-dihydrobenzo b 1,4 dioxine-6-sulfonyl chloride, 2h,3h-benzo 3,4-e 1,4-dioxin-6-ylchlorosulfone, 6-chlorosulfonyl-benzo-1,4-dioxane, 6-chlorosulphonyl-2,3-dihydro-1,4-benzodioxine
2,3-dihydro-1,4-benzodioxine-6-sulfonyl chloride

Specifications

Amber Glass Bottle
63758-12-3
100.0
C8H7ClO4S
1,4-benzodioxan-6-sulfonyl chloride, 1,4-benzodioxin-6-sulfonyl chloride, 2,3-dihydro, 1,4-benzodioxin-6-sulfonylchloride, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxine-6-sulphonylchloride, 2,3-dihydro-benzo 1,4 dioxine-6-sulfonyl chloride, 2,3-dihydro-benzo 1,4 dioxine-6-sulfonylchloride, 2,3-dihydrobenzo b 1,4 dioxine-6-sulfonyl chloride, 2h,3h-benzo 3,4-e 1,4-dioxin-6-ylchlorosulfone, 6-chlorosulfonyl-benzo-1,4-dioxane, 6-chlorosulphonyl-2,3-dihydro-1,4-benzodioxine
C1COC2=C(O1)C=CC(=C2)S(=O)(=O)Cl
234.65
234.66
2, 3-Dihydro-1, 4-benzodioxine-6-sulfonyl chloride
1g
96.5
97%
MFCD02677681
JWHSRWUHRLYAPM-UHFFFAOYSA-N
2,3-dihydro-1,4-benzodioxine-6-sulfonyl chloride
2776169
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS