Please login to your online account to display your discounted pricing

2,3-Dihydro-1,4-benzodioxine-6-sulfonyl chloride, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC01303DE

 View more versions of this product

Catalog No. CC01303DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 234.66
Quantity 5g
Assay 97%
CAS Max % 100.0
CAS 63758-12-3
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
CAS Min % 96.5
MDL Number MFCD02677681
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-6-sulfonyl chloride
Molecular Formula C8H7ClO4S
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-6-sulfonyl chloride
CAS 63758-12-3
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H7ClO4S
Formula Weight 234.66
MDL Number MFCD02677681
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents