missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC01301DE
Specifications
2, 3-Dihydro-1, 4-benzodioxine-6-carboxylic acid | |
94.5 | |
95% | |
MFCD00463509 | |
1,4-benzodioxane-6-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-6-carboxylic acid, 3,4-ethylenedioxy benzoic acid, 2,3-dihydro-benzo 1,4 dioxine-6-carboxylic acid, 1,4-benzodioxin-6-carboxylic acid, 2,3-dihydro, 3,4-ethylenedioxybenzoic acid, 1,4-benzodioxan-6-carboxylic acid, 2,3-dihydro-1,4-benzodioxin-6-carboxylic acid, 2,3-dihydro-1,4-benzodioxine-6-carboxylicacid, 6-carboxy-2,3-dihydro-1,4-benzodioxine | |
C1COC2=C(O1)C=CC(=C2)C(=O)O | |
180.159 | |
180.16 |
4442-54-0 | |
100.0 | |
C9H8O4 | |
5g | |
JWZQJTGQFHIRFQ-UHFFFAOYSA-N | |
2,3-dihydro-1,4-benzodioxine-6-carboxylic acid | |
2758833 | |
95% |