Please login to your online account to display your discounted pricing

2,3-Dihydro-1,4-benzodioxine-6-carbonyl chloride, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  CC01302DA

 View more versions of this product

Catalog No. CC01302DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 198.6
Quantity 1g
Assay 97%
CAS Max % 100.0
Molecular Formula C9H7ClO3
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-6-carbonyl chloride
Assay Percent Range 97%
MDL Number MFCD02681889
CAS 6761-70-2
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-6-carbonyl chloride
CAS 6761-70-2
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7ClO3
Formula Weight 198.6
MDL Number MFCD02681889
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.