missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydro-1,4-benzodioxine-6-carbonyl chloride, 97%, Maybridge

$192.13 - $192.13

Chemical Identifiers

CAS 6761-70-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H7ClO3
Formula Weight 198.6
MDL Number MFCD02681889
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-6-carbonyl chloride
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC01302DA SDS Fisher Scientific Company
CC01302DA
1g Each for $192.13
CC01302CB SDS Fisher Scientific Company
CC01302CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6761-70-2
100.0
C9H7ClO3
MFCD02681889
97%
96.5
97%
198.6
2, 3-Dihydro-1, 4-benzodioxine-6-carbonyl chloride

Specifications

198.6
100.0
MFCD02681889
97%
2, 3-Dihydro-1, 4-benzodioxine-6-carbonyl chloride
1g
C9H7ClO3
96.5
6761-70-2
97%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger