missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dihydro-1,4-benzodioxine-6-carbaldehyde 97%, Maybridge

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C9H8O3
Formula Weight 164.16
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-6-carbaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC01304DA SDS Fisher Scientific Company
CC01304DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC01304DE SDS Fisher Scientific Company
CC01304DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
164.16
97%
C9H8O3
2, 3-Dihydro-1, 4-benzodioxine-6-carbaldehyde

Specifications

1g
Amber glass bottle
C9H8O3
2, 3-Dihydro-1, 4-benzodioxine-6-carbaldehyde
29668-44-8
97%
164.16
97%