missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dihydro-1,4-benzodioxine-6-carbaldehyde 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC01304DA
Specifications
2, 3-Dihydro-1, 4-benzodioxine-6-carbaldehyde | |
97% | |
C9H8O3 | |
1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde | |
C1COC2=C(O1)C=CC(=C2)C=O | |
164.16 | |
164.16 |
29668-44-8 | |
Amber glass bottle | |
1g | |
CWKXDPPQCVWXAG-UHFFFAOYSA-N | |
2,3-dihydro-1,4-benzodioxine-6-carbaldehyde | |
248127 | |
97% |
Chemical Identifiers
29668-44-8 | |
164.16 | |
1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde | |
2,3-dihydro-1,4-benzodioxine-6-carbaldehyde |
C9H8O3 | |
CWKXDPPQCVWXAG-UHFFFAOYSA-N | |
248127 | |
C1COC2=C(O1)C=CC(=C2)C=O |