Please login to your online account to display your discounted pricing

2,3-Dihydro-1,4-benzodioxine-5-carboxamide, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00789CB

 View more versions of this product

Catalog No. MO00789CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 179.17
Quantity 250mg
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
MDL Number MFCD03659722
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-5-carboxamide
Assay Percent Range 97%
Molecular Formula C9H9NO3
CAS 349550-81-8

Specifications

Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-5-carboxamide
CAS 349550-81-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H9NO3
Formula Weight 179.17
MDL Number MFCD03659722