Learn More
2,3-Dihydro-1,4-benzodioxine-5-carbonitrile, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC01416DA
Specifications
2, 3-Dihydro-1, 4-benzodioxine-5-carbonitrile | |
96.5 | |
97% | |
C9H7NO2 | |
1g | |
WNNQCWLSQDNACP-UHFFFAOYSA-N | |
2,3-dihydro-1,4-benzodioxine-5-carbonitrile | |
2795043 | |
97% |
148703-14-4 | |
100.0 | |
Amber Glass Bottle | |
MFCD01116986 | |
2,3-dihydrobenzo b 1,4 dioxine-5-carbonitrile, 1,4-benzodioxin-5-carbonitrile,2,3-dihydro, acmc-1c2ut, 2h,3h-benzo e 1,4-dioxane-5-carbonitrile | |
C1COC2=C(C=CC=C2O1)C#N | |
161.16 | |
161.16 |
Chemical Identifiers
148703-14-4 | |
161.16 | |
WNNQCWLSQDNACP-UHFFFAOYSA-N | |
2795043 | |
C1COC2=C(C=CC=C2O1)C#N |
C9H7NO2 | |
MFCD01116986 | |
2,3-dihydrobenzo b 1,4 dioxine-5-carbonitrile, 1,4-benzodioxin-5-carbonitrile,2,3-dihydro, acmc-1c2ut, 2h,3h-benzo e 1,4-dioxane-5-carbonitrile | |
2,3-dihydro-1,4-benzodioxine-5-carbonitrile |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED.
IF IN EYES.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning