Please login to your online account to display your discounted pricing

2,3-Dihydro-1,4-benzodioxin-6-ylmethanol, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC01309DA

 View more versions of this product

Catalog No. CC01309DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 166.18
Quantity 1g
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
CAS 39270-39-8
MDL Number MFCD01317581
CAS Min % 96.5
Packaging Amber Glass Bottle
Molecular Formula C9H10O3
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxin-6-ylmethanol

Specifications

Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxin-6-ylmethanol
CAS 39270-39-8
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H10O3
Formula Weight 166.18
MDL Number MFCD01317581