missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dihydro-1,4-benzodioxin-6-ylmethanol, Thermo Scientific™
Chemical Identifiers
CAS | 39270-39-8 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.18 |
MDL Number | MFCD01317581 |
InChI Key | FFLHNBGNAWYMRH-UHFFFAOYSA-N |
Synonym | 2,3-dihydrobenzo b 1,4 dioxin-6-yl methanol, 2,3-dihydro-1,4-benzodioxin-6-methanol, 1,4-benzodioxin-6-methanol, 2,3-dihydro, 1,4-benzodioxin-6-methanol,2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-6-yl methanol, 1,4-benzodioxan-6-methanol, 2,3-dihydro-1,4-benzodioxin-6-yl-methanol, 2,3-dihydro-benzo 1,4 dioxin-6-yl methanol, 2,3-dihydrobenzo 1,4 dioxin-6-yl-methanol, 2,3-dihydro-benzo 1,4 dioxin-6-yl-methanol |
PubChem CID | 2776174 |
IUPAC Name | (2,3-dihydro-1,4-benzodioxin-6-yl)methanol |
SMILES | OCC1=CC=C2OCCOC2=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
CC01309CB
|
Thermo Scientific Chemicals
CC01309CB |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC01309DA
|
Thermo Scientific Chemicals
CC01309DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
39270-39-8 | |
166.18 | |
FFLHNBGNAWYMRH-UHFFFAOYSA-N | |
2776174 | |
OCC1=CC=C2OCCOC2=C1 |
C9H10O3 | |
MFCD01317581 | |
2,3-dihydrobenzo b 1,4 dioxin-6-yl methanol, 2,3-dihydro-1,4-benzodioxin-6-methanol, 1,4-benzodioxin-6-methanol, 2,3-dihydro, 1,4-benzodioxin-6-methanol,2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-6-yl methanol, 1,4-benzodioxan-6-methanol, 2,3-dihydro-1,4-benzodioxin-6-yl-methanol, 2,3-dihydro-benzo 1,4 dioxin-6-yl methanol, 2,3-dihydrobenzo 1,4 dioxin-6-yl-methanol, 2,3-dihydro-benzo 1,4 dioxin-6-yl-methanol | |
(2,3-dihydro-1,4-benzodioxin-6-yl)methanol |
Specifications
39270-39-8 | |
100.0 | |
C9H10O3 | |
250mg | |
FFLHNBGNAWYMRH-UHFFFAOYSA-N | |
2,3-dihydro-1,4-benzodioxin-6-ylmethanol | |
2776174 | |
97% |
96.5 | |
97% | |
MFCD01317581 | |
2,3-dihydrobenzo b 1,4 dioxin-6-yl methanol, 2,3-dihydro-1,4-benzodioxin-6-methanol, 1,4-benzodioxin-6-methanol, 2,3-dihydro, 1,4-benzodioxin-6-methanol,2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-6-yl methanol, 1,4-benzodioxan-6-methanol, 2,3-dihydro-1,4-benzodioxin-6-yl-methanol, 2,3-dihydro-benzo 1,4 dioxin-6-yl methanol, 2,3-dihydrobenzo 1,4 dioxin-6-yl-methanol, 2,3-dihydro-benzo 1,4 dioxin-6-yl-methanol | |
OCC1=CC=C2OCCOC2=C1 | |
166.18 | |
166.18 | |
2, 3-Dihydro-1, 4-benzodioxin-6-ylmethanol |