Please login to your online account to display your discounted pricing

2,3-Dihydro-1,4-benzodioxin-5-ylmethylamine, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC01413CB

 View more versions of this product

Catalog No. CC01413CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 201.65
Quantity 250mg
Assay 95%
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxin-5-ylmethylamine hydrochloride
Packaging Amber Glass Bottle
Molecular Formula C9H11NO2·ClH
CAS 261633-71-0
Assay Percent Range 95%

Specifications

Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxin-5-ylmethylamine hydrochloride
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H11NO2·ClH
Formula Weight 201.65