missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Difluoropyridine-4-carboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 851386-31-7
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.09
InChI Key FFGHMEKFPCGYEG-UHFFFAOYSA-N
Synonym 2,3-difluoro-4-carboxypyridine, 2,3-difluoro-4-pyridinecarboxylic acid, 2,3-difluoroisonicotinic acid, 2,3-difluoroisonicotinicacid, 2,3-fifluoroisonicotinic acid, 4-carboxy-2,3-difluoropyridine, 4-pyridinecarboxylic acid, 2,3-difluoro, 5,6-difluoropyridine-4-carboxylic acid, ksc447o0d, pubchem18431
PubChem CID 11240637
IUPAC Name 2,3-difluoropyridine-4-carboxylic acid
SMILES C1=CN=C(C(=C1C(=O)O)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC446770010
SDS View Documents
Thermo Fisher Scientific
AC446770010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC446770050
SDS View Documents
Thermo Fisher Scientific
AC446770050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

851386-31-7
159.09
2,3-difluoro-4-carboxypyridine, 2,3-difluoro-4-pyridinecarboxylic acid, 2,3-difluoroisonicotinic acid, 2,3-difluoroisonicotinicacid, 2,3-fifluoroisonicotinic acid, 4-carboxy-2,3-difluoropyridine, 4-pyridinecarboxylic acid, 2,3-difluoro, 5,6-difluoropyridine-4-carboxylic acid, ksc447o0d, pubchem18431
2,3-difluoropyridine-4-carboxylic acid
C6H3F2NO2
FFGHMEKFPCGYEG-UHFFFAOYSA-N
11240637
C1=CN=C(C(=C1C(=O)O)F)F

Specifications

Glass Bottle
1g
97%
2,3-difluoro-4-carboxypyridine, 2,3-difluoro-4-pyridinecarboxylic acid, 2,3-difluoroisonicotinic acid, 2,3-difluoroisonicotinicacid, 2,3-fifluoroisonicotinic acid, 4-carboxy-2,3-difluoropyridine, 4-pyridinecarboxylic acid, 2,3-difluoro, 5,6-difluoropyridine-4-carboxylic acid, ksc447o0d, pubchem18431
C1=CN=C(C(=C1C(=O)O)F)F
159.09
159.09
2, 3-Difluoropyridine-4-carboxylic acid
165°C to 167°C
851386-31-7
C6H3F2NO2
FFGHMEKFPCGYEG-UHFFFAOYSA-N
2,3-difluoropyridine-4-carboxylic acid
11240637
97%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS