Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',3'-Difluoropropiophenone, 95%

$107.30 - $424.84

Chemical Identifiers

Chemical Name or Material 2',3'-Difluoropropiophenone
CAS 101712-19-0
Molecular Formula C9H8F2O
Formula Weight 170.16
MDL Number MFCD00061141
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2003203

 
alfa aesar™
B2003203
1g Each for $107.30

AAB2003206

 
alfa aesar™
B2003206
5g Each for $424.84
Description & Specifications

Specifications

Formula Weight 170.16
Chemical Name or Material 2',3'-Difluoropropiophenone
Assay 95%
Molecular Formula C9H8F2O
MDL Number MFCD00061141
CAS 101712-19-0
Quantity 1g