missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Difluorophenylboronic acid, 97+%, ACROS Organics™

Chemical Identifiers

CAS 121219-16-7
Molecular Formula C6H5BF2O2
Molecular Weight (g/mol) 157.91
MDL Number MFCD01863170
InChI Key SZYXKFKWFYUOGZ-UHFFFAOYSA-N
Synonym 2,3-difluoro phenylboric acid
PubChem CID 2734333
IUPAC Name (2,3-difluorophenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)F)F)(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC359120010 SDS View Documents Fisher Scientific Acros
359120010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

121219-16-7
157.91
SZYXKFKWFYUOGZ-UHFFFAOYSA-N
2734333
B(C1=C(C(=CC=C1)F)F)(O)O
C6H5BF2O2
MFCD01863170
2,3-difluoro phenylboric acid
(2,3-difluorophenyl)boronic acid

Specifications

Authentic
White to Beige
121219-16-7
100.0
C6H5BF2O2
2,3-difluoro phenylboric acid
B(C1=C(C(=CC=C1)F)F)(O)O
157.91
157.91
97+%
Glass bottle
1g
97.0
97% min. (HPLC)
MFCD01863170
SZYXKFKWFYUOGZ-UHFFFAOYSA-N
(2,3-difluorophenyl)boronic acid
2734333
Crystalline Powder
2, 3-Difluorophenylboronic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning