Please login to your online account to display your discounted pricing

2,3-Difluorophenylboronic acid, 97+%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  35912-0100

 View more versions of this product

Catalog No. AC359120100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.0%
Physical Form Crystalline Powder
Formula Weight 157.91
Quantity 10g
Color White to light beige
CAS Max % 100.0
CAS Min % 97.0
Infrared Spectrum Authentic
Molecular Formula C6H5BF2O2
CAS 121219-16-7
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD01863170
Packaging Glass bottle
Assay Percent Range 97+%
Chemical Name or Material 2, 3-Difluorophenylboronic acid
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 2, 3-Difluorophenylboronic acid
CAS 121219-16-7
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C6H5BF2O2
Formula Weight 157.91
MDL Number MFCD01863170
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents