missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Difluorophenylacetic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC435470050
Specifications
2, 3-Difluorophenylacetic acid | |
White | |
5g | |
96.0 | |
97% min. (HPLC) | |
MFCD00040968 | |
UXSQXUSJGPVOKT-UHFFFAOYSA-N | |
2-(2,3-difluorophenyl)acetic acid | |
520772 | |
Crystalline Powder |
Authentic | |
117.0°C to 119.0°C | |
145689-41-4 | |
100.0 | |
C8H6F2O2 | |
2,3-difluorobenzeneacetic acid | |
C1=CC(=C(C(=C1)F)F)CC(=O)O | |
172.13 | |
172.13 | |
97% |
Chemical Identifiers
145689-41-4 | |
172.13 | |
UXSQXUSJGPVOKT-UHFFFAOYSA-N | |
520772 | |
C1=CC(=C(C(=C1)F)F)CC(=O)O |
C8H6F2O2 | |
MFCD00040968 | |
2,3-difluorobenzeneacetic acid | |
2-(2,3-difluorophenyl)acetic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Warning