missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Difluorophenylacetic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 145689-41-4
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD00040968
Chemical Name or Material 2, 3-Difluorophenylacetic acid
Assay ≥97.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC435470010 SDS Thermo Fisher Scientific
AC435470010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC435470050 SDS Thermo Fisher Scientific
AC435470050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

145689-41-4
100.0
C8H6F2O2
MFCD00040968
≥97.0%
96.0
97%
172.13
2, 3-Difluorophenylacetic acid

Specifications

Crystalline Powder
1g
116° to 120°C
96.0
97%
C8H6F2O2
2, 3-Difluorophenylacetic acid
172.13
White to off-white
100.0
Authentic
145689-41-4
MFCD00040968
≥97.0%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.