Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluorocinnamic acid, 98%

$351.08 - $351.08

Chemical Identifiers

Chemical Name or Material 2,3-Difluorocinnamic acid
CAS 207981-48-4
Molecular Formula C9H6F2O2
Formula Weight 184.14
MDL Number MFCD00061291
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2010506

  SDS
alfa aesar™
B2010506
5g Each for $351.08
Description & Specifications

Specifications

Formula Weight 184.14
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Assay 98%
MDL Number MFCD00061291
Chemical Name or Material 2,3-Difluorocinnamic acid
Health Hazard 1 Warning
Melting Point 198° to 201°C
CAS 207981-48-4
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C9H6F2O2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.