Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluorobenzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 2,3-Difluorobenzyl bromide
CAS 113211-94-2
Molecular Formula C7H5BrF2
Formula Weight 207.02
MDL Number MFCD00042488
UN Number UN1760
Beilstein 7089244
Synonym alpha-Bromo-2,3-difluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2021403

  SDS
alfa aesar™
B2021403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2021406

  SDS
alfa aesar™
B2021406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2021414

  SDS
alfa aesar™
B2021414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 207.02
Chemical Name or Material 2,3-Difluorobenzyl bromide
Assay 97%
Molecular Formula C7H5BrF2
MDL Number MFCD00042488
CAS 113211-94-2
UN Number UN1760
Beilstein 7089244
Synonym alpha-Bromo-2,3-difluorotoluene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
Flash Point 90°C (194°F)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.