Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluorobenzophenone, 97%

$165.74 - $165.74

Chemical Identifiers

Chemical Name or Material 2,3-Difluorobenzophenone
CAS 208173-20-0
Molecular Formula C13H8F2O
Formula Weight 218.2
MDL Number MFCD00061139
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1995106

  SDS
alfa aesar™
A1995106
5g Each for $165.74
Description & Specifications

Specifications

Formula Weight 218.2
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 1 Warning
MDL Number MFCD00061139
CAS 208173-20-0
Assay 97%
Chemical Name or Material 2,3-Difluorobenzophenone
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 176° to 178°C (16mmHg)
Molecular Formula C13H8F2O
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.