Please login to your online account to display your discounted pricing

2,3-Difluorobenzoic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3-Difluorobenzoic acid
Name Note 98%
CAS 4519-39-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4F2O2
Linear Formula F2C6H3COOH
Formula Weight 158.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC311470010

  SDS
thermo fisher scientific
AC311470010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC311470100

  SDS
thermo fisher scientific
311470100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 158.11
Quantity 1g
Color White to off-white
TSCA TSCA
Melting Point 161° to 165°C
CAS Max % 100.0
Chemical Name or Material 2, 3-Difluorobenzoic acid
Assay Percent Range 98%
Molecular Formula C7H4F2O2
Linear Formula F2C6H3COOH
MDL Number MFCD00010267
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Name Note 98%
CAS 4519-39-5
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.