missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6264903

 View more versions of this product

Catalog No. AAH6264903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2,3-Difluorobenzeneboronic acid pinacol ester
1073339-17-9
2-2,3-difluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2,3-difluorophenylboronic acid pinacol ester, 2,3-difluorobenzeneboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-2,3-difluorophenyl-4,4,5,5-tetramethyl
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)F)F
240.057
240.05
96%
1g
C12H15BF2O2
BFCCQFHEIXEEKP-UHFFFAOYSA-N
2-(2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
56973143
Solid

Chemical Identifiers

1073339-17-9
240.057
2-2,3-difluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2,3-difluorophenylboronic acid pinacol ester, 2,3-difluorobenzeneboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-2,3-difluorophenyl-4,4,5,5-tetramethyl
2-(2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C12H15BF2O2
BFCCQFHEIXEEKP-UHFFFAOYSA-N
56973143
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)F)F
Documents