Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluorobenzeneboronic acid, 98%

$121.74 - $421.76

Chemical Identifiers

Chemical Name or Material 2,3-Difluorobenzeneboronic acid
CAS 121219-16-7
Molecular Formula C6H5BF2O2
Formula Weight 157.91
MDL Number MFCD01863170
Beilstein 6594341
Synonym 2,3-Difluorophenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1801206

 
alfa aesar™
L1801206
5g Each for $121.74

AAL1801214

 
alfa aesar™
L1801214
25g Each for $421.76
Description & Specifications

Specifications

Formula Weight 157.91
Chemical Name or Material 2,3-Difluorobenzeneboronic acid
MDL Number MFCD01863170
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C6H5BF2O2
CAS 121219-16-7
Beilstein 6594341
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈225°C
Synonym 2,3-Difluorophenylboronic acid
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.