missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Difluorobenzaldehyde, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC303510050

 View more versions of this product

Catalog No. AC303510050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 3-Difluorobenzaldehyde
Clear pale yellow
2646-91-5
100.0
Glass bottle
Liquid
98%
FC6H3CHO
MFCD00010292
1.3g/mL
5g
97.5
Authentic
1.4980 to 1.5000 (20°C, 589nm)
≥97.5%
C7H4F2O
142.11

Chemical Identifiers

2646-91-5
100.0
C7H4F2O
142.11
2, 3-Difluorobenzaldehyde
97.5
98%
FC6H3CHO
MFCD00010292
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning