Please login to your online account to display your discounted pricing

2,3-Difluorobenzaldehyde, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3-Difluorobenzaldehyde
CAS 2646-91-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4F2O
Linear Formula FC6H3CHO
Formula Weight 142.11
MDL Number MFCD00010292
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC303510050

  SDS
thermo fisher scientific
AC303510050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC303510010

  SDS
thermo fisher scientific
303510010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 142.11
Assay ≥97.5%
Quantity 5g
Color Clear pale yellow
Density 1.3g/mL
CAS Max % 100.0
Refractive Index 1.4980 to 1.5000 (20°C, 589nm)
CAS 2646-91-5
Molecular Formula C7H4F2O
Infrared Spectrum Authentic
Assay Percent Range 98%
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00010292
Packaging Glass bottle
CAS Min % 97.5
Linear Formula FC6H3CHO
Chemical Name or Material 2, 3-Difluorobenzaldehyde
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.