Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoroanisole, 97+%

$84.04 - $84.04

Chemical Identifiers

Chemical Name or Material 2,3-Difluoroanisole
CAS 134364-69-5
Molecular Formula C7H6F2O
Formula Weight 144.12
MDL Number MFCD01631562
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2291603

  SDS
alfa aesar™
B2291603
1g Each for $84.04
Description & Specifications

Specifications

Formula Weight 144.12
Chemical Name or Material 2,3-Difluoroanisole
Assay ≥97%
Molecular Formula C7H6F2O
MDL Number MFCD01631562
CAS 134364-69-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 62°C (144°F)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.