Please login to your online account to display your discounted pricing

2,3-Difluoroaniline, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3-Difluoroaniline
CAS 4519-40-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H5F2N
Linear Formula F2C6H3NH2
Formula Weight 129.11
MDL Number MFCD00010298
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC303540010

  SDS
thermo fisher scientific
303540010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 129.11
Physical Form Liquid
Quantity 1g
Color Clear light orange
Density 1.2740g/mL
CAS Max % 100.0
Linear Formula F2C6H3NH2
Synonym 2, 3-Difluorobenzenamine
CAS 4519-40-8
Molecular Formula C6H5F2N
Assay Percent Range 98%
MDL Number MFCD00010298
CAS Min % 97.5
Storage Information Product darkens on storage
Refractive Index 1.5120 to 1.5140 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material 2, 3-Difluoroaniline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.