Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoroaniline, 98%

Chemical Identifiers

Chemical Name or Material 2,3-Difluoroaniline
CAS 4519-40-8
Molecular Formula C6H5F2N
Formula Weight 129.11
MDL Number MFCD00010298
UN Number UN2941
Beilstein 2716500
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2166503

  SDS
alfa aesar™
B2166503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 129.11
MDL Number MFCD00010298
Beilstein 2716500
Flash Point 71°C (159°F)
Chemical Name or Material 2,3-Difluoroaniline
Assay 98%
UN Number UN2941
CAS 4519-40-8
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Molecular Formula C6H5F2N
Health Hazard 1 Danger
Boiling Point 68° to 69°C (12mmHg)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES
Odor Characteristic
Quantity 1g
Health Hazard 2 GHS H Statement
H311-H331-H373-H227-H302-H315-H319
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H331-H373-H227-H302-H315-H319
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.