Please login to your online account to display your discounted pricing

2,3-Difluoro-6-nitrophenol, 97%, ACROS Organics™

$536.34 - $536.34

Chemical Identifiers

Chemical Name or Material 2, 3-Difluoro-6-nitrophenol
Name Note 97%
CAS 82419-26-9
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6 H3 F2 N O3
Linear Formula F2C6H2(NO2)OH
Formula Weight 175.09
MDL Number MFCD00042255
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC336320010

  SDS
thermo fisher scientific
AC336320010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC336320250

  SDS
thermo fisher scientific
336320250
Glass bottle 25g Each for $536.34
Description & Specifications

Specifications

Formula Weight 175.09
Quantity 1g
TSCA TSCA
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
CAS 82419-26-9
Chemical Name or Material 2, 3-Difluoro-6-nitrophenol
Molecular Formula C6 H3 F2 N O3
Assay Percent Range 97%
Linear Formula F2C6H2(NO2)OH
Name Note 97%
MDL Number MFCD00042255