Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoro-6-nitrophenol, 97%

$47.46 - $47.46

Chemical Identifiers

Chemical Name or Material 2,3-Difluoro-6-nitrophenol
CAS 82419-26-9
Molecular Formula C6H3F2NO3
Formula Weight 175.09
MDL Number MFCD00042255
Beilstein 4410368
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2296503

  SDS
alfa aesar™
B2296503
1g Each for $47.46
Description & Specifications

Specifications

Formula Weight 175.09
Assay 97%
Quantity 1g
Chemical Name or Material 2,3-Difluoro-6-nitrophenol
Health Hazard 1 Danger
CAS 82419-26-9
Melting Point 60° to 62°C
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 4410368
MDL Number MFCD00042255
Molecular Formula C6H3F2NO3
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.