missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoro-6-methylpyridine, 98%

$29.45 - $345.43

Chemical Identifiers

CAS 1227579-04-5
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD16610828
InChI Key HGBWAUANMSZOIJ-UHFFFAOYSA-N
Synonym 2,3-difluoro-6-methyl-pyridine
PubChem CID 70680696
IUPAC Name 2,3-difluoro-6-methylpyridine
SMILES CC1=NC(=C(C=C1)F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64514MD View Documents Alfa Aesar™
H64514MD
250mg Each for $29.45
 
AAH6451403 View Documents Alfa Aesar™
H6451403
1g Each for $86.99
 
AAH6451406 View Documents Alfa Aesar™
H6451406
5g Each for $345.43
 
Specifications

Chemical Identifiers

1227579-04-5
129.11
HGBWAUANMSZOIJ-UHFFFAOYSA-N
70680696
CC1=NC(=C(C=C1)F)F
C6H5F2N
MFCD16610828
2,3-difluoro-6-methyl-pyridine
2,3-difluoro-6-methylpyridine

Specifications

Colorless to Yellow
1227579-04-5
MFCD16610828
HGBWAUANMSZOIJ-UHFFFAOYSA-N
2,3-difluoro-6-methylpyridine
70680696
Liquid
2,3-Difluoro-6-methylpyridine
250mg
C6H5F2N
2,3-difluoro-6-methyl-pyridine
CC1=NC(=C(C=C1)F)F
129.11
129.11
98%