Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoro-6-methoxyphenylacetic acid, 97%

$209.34 - $209.34

Chemical Identifiers

Chemical Name or Material 2,3-Difluoro-6-methoxyphenylacetic acid
Molecular Formula C9H8F2O3
Formula Weight 202.16
MDL Number MFCD04116025
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2677903

 
alfa aesar™
H2677903
1g Each for $209.34

AAH26779MD

 
alfa aesar™
H26779MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 202.16
Chemical Name or Material 2,3-Difluoro-6-methoxyphenylacetic acid
Assay 97%
Molecular Formula C9H8F2O3
MDL Number MFCD04116025
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.