Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoro-6-methoxyphenol, 97%

$190.86 - $190.86

Chemical Identifiers

Chemical Name or Material 2,3-Difluoro-6-methoxyphenol
CAS 186306-70-7
Molecular Formula C7H6F2O2
Formula Weight 160.12
MDL Number MFCD04116024
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2645903

  SDS
alfa aesar™
H2645903
1g Each for $190.86
Description & Specifications

Specifications

Formula Weight 160.12
Molecular Formula C7H6F2O2
MDL Number MFCD04116024
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
Melting Point 44° to 47°C
Chemical Name or Material 2,3-Difluoro-6-methoxyphenol
Assay 97%
CAS 186306-70-7
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.