Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Difluoro-6-methoxybenzylamine, 97%

Chemical Identifiers

Chemical Name or Material 2,3-Difluoro-6-methoxybenzylamine
CAS 886501-77-5
Molecular Formula C8H9F2NO
Formula Weight 173.16
MDL Number MFCD04116022
UN Number UN2735
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2667003

  SDS
alfa aesar™
H2667003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH26670MD

  SDS
alfa aesar™
H26670MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.16
Molecular Formula C8H9F2NO
Chemical Name or Material 2,3-Difluoro-6-methoxybenzylamine
MDL Number MFCD04116022
Health Hazard 1 Danger
Quantity 1g
UN Number UN2735
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
CAS 886501-77-5
Assay 97%
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314
Causes severe skin burns and eye damage.