Learn More
2,3-Difluoro-6-methoxybenzyl bromide, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H2638006
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2,3-Difluoro-6-methoxybenzyl bromide | |
46°C to 48°C | |
MFCD04116023 | |
UN3261 | |
SVUCRCMGEHPJGQ-UHFFFAOYSA-N | |
3-(bromomethyl)-1,2-difluoro-4-methoxybenzene | |
4995861 | |
97% |
886501-83-3 | |
C8H7BrF2O | |
5 g | |
2,3-difluoro-6-methoxybenzyl bromide, 2-bromomethyl-3,4-difluoro-1-methoxybenzene, 2,3-difluoro-6-methoxybenzyl-bromide, 2,3-difluoro-6-methoxy-benzyl bromide, 3-bromomethyl-1,2-difluoro-4-methoxybenzene, benzene,2-bromomethyl-3,4-difluoro-1-methoxy | |
COC1=C(CBr)C(F)=C(F)C=C1 | |
237.04 | |
237.04 |
Chemical Identifiers
886501-83-3 | |
237.04 | |
SVUCRCMGEHPJGQ-UHFFFAOYSA-N | |
4995861 | |
COC1=C(CBr)C(F)=C(F)C=C1 |
C8H7BrF2O | |
MFCD04116023 | |
2,3-difluoro-6-methoxybenzyl bromide, 2-bromomethyl-3,4-difluoro-1-methoxybenzene, 2,3-difluoro-6-methoxybenzyl-bromide, 2,3-difluoro-6-methoxy-benzyl bromide, 3-bromomethyl-1,2-difluoro-4-methoxybenzene, benzene,2-bromomethyl-3,4-difluoro-1-methoxy | |
3-(bromomethyl)-1,2-difluoro-4-methoxybenzene |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures