missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Difluoro-4-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC446780050

 View more versions of this product

Catalog No. AC446780050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 3-Difluoro-4-(trifluoromethyl)pyridine
5g
96% min. (GC)
2,3-difluoro-4-trifluoromethyl pyridine, alpha,alpha,alpha,2,3-pentafluoro-4-picoline
C1=CN=C(C(=C1C(F)(F)F)F)F
183.07
183.07
82.0°C to 84.0°C (400.0 Torr)
1215034-77-7
C6H2F5N
ZOMVJIOHXSXCIP-UHFFFAOYSA-N
2,3-difluoro-4-(trifluoromethyl)pyridine
13491638
97%

Chemical Identifiers

1215034-77-7
183.07
2,3-difluoro-4-trifluoromethyl pyridine, alpha,alpha,alpha,2,3-pentafluoro-4-picoline
2,3-difluoro-4-(trifluoromethyl)pyridine
C6H2F5N
ZOMVJIOHXSXCIP-UHFFFAOYSA-N
13491638
C1=CN=C(C(=C1C(F)(F)F)F)F
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapour.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning

Documents